Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Thermo Scientific Chemicals
Oxonic acid, potassium salt, 97.5%
CAS: 2207-75-2 | C4H2KN3O4 | 195.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA168530250
Price (MYR)
999.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
999.00
Add to cart
Chemical Identifiers
CAS60-23-1
IUPAC Name2-aminoethane-1-thiol
Molecular FormulaC2H7NS
InChI KeyUFULAYFCSOUIOV-UHFFFAOYSA-N
SMILESNCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration Iodimetric>=94 %
Appearance (Form)Powder or crystals
Melting point94°C to 99°C