2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA168365000
Price (MYR)
963.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
963.00
Add to cart
Chemical Identifiers
CAS19872-52-7
IUPAC Name4-methyl-4-sulfanylpentan-2-one
Molecular FormulaC6H12OS
InChI KeyQRNZMFDCKKEPSX-UHFFFAOYSA-N
SMILESCC(=O)CC(C)(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4600-1.4640 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%