2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA168361000
Price (MYR)
348.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
348.00
Add to cart
Chemical Identifiers
CAS912350-00-6
IUPAC Namepotassium trifluoro(4-iodophenyl)boranuide
Molecular FormulaC6H4BF3IK
InChI KeyWTXNSBOVCAXUGV-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Melting Point (clear melt)293.0-303.0?C