2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
1 kg
Packaging:
View all
Metal drum
Catalog number FSA168360025
Price (MYR)
1,999.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
1,999.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methyltetrahydrofuran
Melting Point-136.0°C
CAS96-47-9
ColorColorless
Health Hazard 1GHS Signal Word: Danger