2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals
CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA168360010
Price (MYR)
1,339.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,339.00
Add to cart
Chemical Identifiers
CAS84-65-1
IUPAC Name9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O2
InChI KeyRZVHIXYEVGDQDX-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)Yellow-green to tan to khaki
Melting point283°C to 288°C