2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
Catalog number FSA167910025
Price (MYR)
1,002.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,002.00
Add to cart
Chemical Identifiers
CAS1066-54-2
IUPAC Nameethynyltrimethylsilane
Molecular FormulaC5H10Si
InChI KeyCWMFRHBXRUITQE-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3873 to 1.3903 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
For UV/VIS spectroscopy
RUO – Research Use Only