N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA167795000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12672-27-4
IUPAC Namedialuminium(3+) bis(λ²-cobalt(2+)) pentaoxidandiide
Molecular FormulaAl2Co2O5
InChI KeyCRHLEZORXKQUEI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Co++].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofACobalt (Co): 39.0-42.0%
Particle SizeParticle Size (Micro_%90): ≤74 micron (-200 mesh)
Assay from Supplier's CofAAluminum (Al): 24.0-26.0%
For UV/VIS spectroscopy
RUO – Research Use Only