Cyclohexane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Cyclohexane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Cyclohexane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Cyclohexane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexane, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA167740010
Price (MYR)
436.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexane
Name Note99+%
Absorbance0.01 max. at 260nm, 0.02 max. at 250nm, 0.09 max. at 235nm, 0.3 max. at 220nm, 1 max. at 210nm
CAS110-82-7
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only