n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl acetate, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA167680010
Price (MYR)
482.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
482.00
Add to cart
Chemical Identifiers
CAS26444-49-5
IUPAC Name4-methylphenyl diphenyl phosphate
Molecular FormulaC19H17O4P
InChI KeyOJUZRFGUKHQNJX-UHFFFAOYSA-N
SMILESCC1=CC=C(OP(=O)(OC2=CC=CC=C2)OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5580-1.5640 @ 25°C
Appearance (Color)Clear colorless to pale yellow or pale orange
Assay from Supplier's CofA≥92.0% (GC) (Sum of isomers)
For UV/VIS spectroscopy
RUO – Research Use Only