Lauric acid, 99%
Lauric acid, 99%
Lauric acid, 99%
Thermo Scientific Chemicals
Lauric acid, 99%
CAS: 143-07-7 | C12H24O2 | 200.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA167285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS100-39-0
IUPAC Name(bromomethyl)benzene
Molecular FormulaC7H7Br
InChI KeyAGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5745 to 1.5765 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow