Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA167270010
Price (MYR)
1,394.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,394.00
Add to cart
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with CHA>=98.5 % (On dry substance)
Water10.0 to 13.0 % (KF)
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration with CHA=<0.5 % Sulfuric acid