N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, ACROS Organics, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 93 deg.C to 94 deg.C (13.0mmHg), Colorless to Yellow, Melting Point: -42 deg.C, Molecular Weight: 140.26, Percent Purity: 97%, Assay Percent Range: 96% min. (GC), CAS: 18156-74-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA167131000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1676-63-7
IUPAC Name1-(4-ethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O2
InChI KeyYJFNFQHMQJCPRG-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystalline powder, crystals and/or chunks
Assay (GC)≥97.5%
Melting Point (clear melt)34-38?C
Silylation reagent
RUO – Research Use Only