4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA166965000
Price (MYR)
884.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
884.00
Add to cart
Chemical Identifiers
CAS5422-88-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Assay (GC)≥95.0%
Refractive Index1.5415-1.5475 @ 20?C