4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA166961000
Price (MYR)
318.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Chemical Identifiers
CAS78-78-4
IUPAC Name2-methylbutane
Molecular FormulaC5H12
InChI KeyQWTDNUCVQCZILF-UHFFFAOYSA-N
SMILESCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UV1 cm cell vs water
UVat 220 nm A: =<0.07
UVat 240 nm A: =<0.01
Acidity=<0.0005 meq/g