1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA166711000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS507-09-5
IUPAC Nameethanethioic S-acid
Molecular FormulaC2H4OS
InChI KeyDUYAAUVXQSMXQP-UHFFFAOYSA-N
SMILESCC(S)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4610 to 1.4650 (20°C, 589 nm)
Titration Argentometric>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid