Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals
Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals
Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals
Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine-d5, for NMR, 99% atom D, Thermo Scientific Chemicals
CAS: 7291-22-7 | C5H5N | 84.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass Bottle
Catalog number FSA166570250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1129-26-6
IUPAC Name4-methoxybenzene-1-sulfonamide
Molecular FormulaC7H9NO3S
InChI KeyMSFQEZBRFPAFEX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)110-114?C
FormCrystalline powder or solid
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
For NMR
RUO – Research Use Only