Taurine, 99%, Thermo Scientific Chemicals
Taurine, 99%, Thermo Scientific Chemicals
Taurine, 99%, Thermo Scientific Chemicals
Taurine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Taurine, 99%, Thermo Scientific Chemicals
CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA166545000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1758-25-4
IUPAC Name2-(2,5-dimethoxyphenyl)acetic acid
Molecular FormulaC10H12O4
InChI KeyBBZDYQUXRFATHZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(CC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)121.0-128.0?C
Appearance (Color)White to cream to yellow
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5%