Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.5% atom D, Thermo Scientific Chemicals
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
10 mL
Packaging:
Glass Bottle
Catalog number FSA166350500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS106-92-3
IUPAC Name2-[(prop-2-en-1-yloxy)methyl]oxirane
Molecular FormulaC6H10O2
InChI KeyLSWYGACWGAICNM-UHFFFAOYNA-N
SMILESC=CCOCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.2 % (K.F.)
Refractive index1.4332 to 1.4352 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
For NMR
RUO – Research Use Only