Deuterium oxide, for NMR, 100.0% atom D, Thermo Scientific Chemicals
Deuterium oxide, for NMR, 100.0% atom D, Thermo Scientific Chemicals
Deuterium oxide, for NMR, 100.0% atom D, Thermo Scientific Chemicals
Deuterium oxide, for NMR, 100.0% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Deuterium oxide, for NMR, 100.0% atom D, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
10 mL
50 mL
Packaging:
Glass Bottle
Catalog number FSA166311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS629-50-5
IUPAC Nametridecane
Molecular FormulaC13H28
InChI KeyIIYFAKIEWZDVMP-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
For NMR
RUO – Research Use Only