Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, 99.9% atom D, Thermo Scientific Chemicals
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
10 mL
50 mL
Packaging:
Glass Bottle
Catalog number FSA166291000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS108-87-2
IUPAC Namemethylcyclohexane
Molecular FormulaC7H14
InChI KeyUAEPNZWRGJTJPN-UHFFFAOYSA-N
SMILESCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4215-1.4245 @ 20°C (non-U.S. specification)
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms (non-U.S. specification)
For NMR
RUO – Research Use Only