Acetone-d6, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Acetone-d6, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Acetone-d6, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Acetone-d6, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Acetone-d6, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
CAS: 666-52-4 | C3H6O | 64.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
25 mL
Packaging:
Glass Bottle
Catalog number FSA166220500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetone-d6
Density0.8700g/mL
Deuterium (D)99.4% min.
Flash Point−20°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only