Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
25 mL
Catalog number FSA166210500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic-d3 acid-d
Density1.1300g/mL
Deuterium (D)99.4% min.
Flash Point40°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only