Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals
Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals
Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals
Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid-d, for NMR, 98% atom D, Thermo Scientific Chemicals
CAS: 758-12-3 | C2H4O2 | 61.058 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass Bottle
Catalog number FSA166202500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid-d
Density1.0500g/mL
Deuterium (D)97.5% min.
Flash Point40°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only