Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA165895000
Price (MYR)
716.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
716.00
Add to cart
Chemical Identifiers
CAS26878-35-3
IUPAC Namedihydrogen pyridine-2,5-diamine dichloride
Molecular FormulaC5H9Cl2N3
InChI KeyRWHNUWWUPZKDQP-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple to brown
Water Content (Karl Fischer Titration)< 1.5%
Assay (Titration ex Chloride)> 96.0%