Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA165891000
Price (MYR)
305.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
305.00
Add to cart
Chemical Identifiers
CAS1198-30-7
IUPAC Nameisoquinoline-1-carbonitrile
Molecular FormulaC10H6N2
InChI KeyHJHXYSBRTVFEDD-UHFFFAOYSA-N
SMILESN#CC1=C2C=CC=CC2=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or flakes