Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA165082500
Price (MYR)
344.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
344.00
Add to cart
Chemical Identifiers
CAS2491-17-0
IUPAC Name4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
Molecular FormulaC21H33N3O4S
InChI KeyGBCAVSYHPPARHX-UHFFFAOYSA-M
SMILESCC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Melting point112°C to 120°C
HPLC>=94 %
Appearance (Color)White to light yellow