Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number FSA165080010
Price (MYR)
1,013.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,013.00
Add to cart
Specifications
Chemical Name or MaterialPhenyl salicylate
CAS118-55-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.