Ethyl caproate, 99+%, Thermo Scientific Chemicals
Ethyl caproate, 99+%, Thermo Scientific Chemicals
Ethyl caproate, 99+%, Thermo Scientific Chemicals
Ethyl caproate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl caproate, 99+%, Thermo Scientific Chemicals
CAS: 123-66-0 | C8H16O2 | 144.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA164990050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl caproate
Acidity1mg KOH/g max.
CAS123-66-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.