2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
CAS: 75-30-9 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA164961000
Price (MYR)
645.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
645.00
Add to cart
Specifications
Chemical Name or Material2-Iodopropane
Name Note98+%
CAS75-30-9
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning