Methyl isobutyrate, 99%, Thermo Scientific Chemicals
Methyl isobutyrate, 99%, Thermo Scientific Chemicals
Methyl isobutyrate, 99%, Thermo Scientific Chemicals
Methyl isobutyrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl isobutyrate, 99%, Thermo Scientific Chemicals
CAS: 547-63-7 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA164605000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS642-31-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green
FormPowder
Assay (GC)≥97.5%
Free acid (titration)≤1%
Melting Point (clear melt)102.0-108.0?C