4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
CAS: 90-94-8 | C17H20N2O | 268.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA164490050
Price (MYR)
237.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
237.00
Add to cart
Chemical Identifiers
CAS60-35-5
IUPAC Nameacetamide
Molecular FormulaC2H5NO
InChI KeyDLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILESCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)White
Infrared spectrumConforms
Melting point78°C to 81°C
GC>=98.5 %