Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals
CAS: 79-10-7 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA164252500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS528-50-7
IUPAC Name2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+33° to +35° (20°C, 589 nm) (c=4, water)