Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
Packaging:
View all
Plastic Bottle
Catalog number FSA164240050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS1139-30-6
IUPAC Name4,12,12-trimethyl-9-methylidene-5-oxatricyclo[8.2.0.0⁴,⁶]dodecane
Molecular FormulaC15H24O
InChI KeyNVEQFIOZRFFVFW-UHFFFAOYNA-N
SMILESCC12CCC3C(CC3(C)C)C(=C)CCC1O2
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Specific optical rotation-55° to -75° (20°C, 589 nm) (c=10, EtOH)
Appearance (Color)White
Infrared spectrumConforms
Melting point55°C to 64°C