5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
Thermo Scientific Chemicals
5-Methoxysalicylic acid, 98%
CAS: 2612-02-4 | C8H8O4 | 168.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA16387010010 gGlass bottle
Catalog number FSA163870100
Price (MYR)
325.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
325.00
Add to cart
Chemical Identifiers
CAS7782-82-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental Analysis(Selenium, Se) : 49.69-54.91% (Theory 52.31%)
Appearance (Color)White to pale cream to pale gray
Assay (unspecified)(By Selenium Elemental Analysis) : >95.0 to <105.0%