N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
CAS: 91-40-7 | C13H11NO2 | 213.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Packaging:
Plastic bottle
Catalog number FSA163005000
Price (MYR)
2,435.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,435.00
Add to cart
Chemical Identifiers
CAS97-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to orange to black
Assay (GC)≥97.5%
Melting Point (clear melt)46.0-53.0?C
FormCrystals or powder or crystalline powder or lumps or fused solid or flakes
Identification (FTIR)Conforms
Indicator
RUO – Research Use Only