N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Phenylanthranilic acid, 98%, Thermo Scientific Chemicals
CAS: 91-40-7 | C13H11NO2 | 213.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
500 g
Packaging:
Plastic bottle
Catalog number FSA163001000
Price (MYR)
805.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
805.00
Add to cart
Chemical Identifiers
CAS4064-06-6
IUPAC Name{4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-8-yl}methanol
Molecular FormulaC12H20O6
InChI KeyPOORJMIIHXHXAV-UHFFFAOYNA-N
SMILESCC1(C)OC2OC(CO)C3OC(C)(C)OC3C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormViscous liquid or paste
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Optical Rotation-58? ±4? (c =3 in Chloroform)
View more
Indicator
RUO – Research Use Only