Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
CAS: 110-99-6 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
Packaging:
Plastic bottle
Catalog number FSA162260025
Price (MYR)
6,651.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
6,651.00
Add to cart
Chemical Identifiers
CAS2758-18-1
IUPAC Name3-methylcyclopent-2-en-1-one
Molecular FormulaC6H8O
InChI KeyCHCCBPDEADMNCI-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer0.1 % hydroquinone
Appearance (Color)Clear light yellow to yellow-brown
Refractive index1.4870 to 1.4900 (20°C, 589 nm)
Appearance (Form)Liquid