2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
CAS: 108-75-8 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA161955000
Price (MYR)
1,411.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,411.00
Add to cart
Chemical Identifiers
CAS15212-03-0
IUPAC Name2-methoxyphenyl acetate
Molecular FormulaC9H10O3
InChI KeyBHJHPYFAYGAPLS-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5120-1.5160 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless