2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
CAS: 108-75-8 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number FSA161951000
Price (MYR)
556.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Chemical Identifiers
CAS98-58-8
IUPAC Name4-bromobenzene-1-sulfonyl chloride
Molecular FormulaC6H4BrClO2S
InChI KeyKMMHZIBWCXYAAH-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)72.0-78.0?C
Appearance (Color)White to cream to pale brown