N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Catalog number FSA161250250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7783-96-2
IUPAC Namesilver(1+) iodide
Molecular FormulaAgI
InChI KeyMSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES[Ag+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Alkylation reagent
RUO – Research Use Only