Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Phase transfer catalyst
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA161072500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Nitrogen (N)11.5 to 12.8 %
Sulfated ash=<0.1 %
Peroxides=<400 ppm (Titanyl sulphate method)
Appearance of solution(5 % in water at 25°C) Free of haze
Additional infoEuropean colour test - BY colour: > BY6
View more
Phase transfer catalyst
RUO – Research Use Only