3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA161061000
Price (MYR)
679.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
679.00
Add to cart
Chemical Identifiers
CAS1305-79-9
IUPAC Namecalcium dioxidanediide
Molecular FormulaCaO2
InChI KeyLHJQIRIGXXHNLA-UHFFFAOYSA-N
SMILES[Ca++].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)65% min