Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 103-72-0 | C7H5NS | 135.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA160971000
Price (MYR)
516.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
516.00
Add to cart
Chemical Identifiers
CAS144-48-9
IUPAC Name2-iodoacetamide
Molecular FormulaC2H4INO
InChI KeyPGLTVOMIXTUURA-UHFFFAOYSA-N
SMILESNC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Iodide)≥97.5 to ≤102.5% (Dry wt. basis)
Assay (HPLC)≥97.5%