n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA160780251
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS6728-26-3
IUPAC Name(2E)-hex-2-enal
Molecular FormulaC6H10O
InChI KeyMBDOYVRWFFCFHM-SNAWJCMRSA-N
SMILESCCCC=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4440 to 1.4500 (20°C, 589 nm)
Density(20°C) 0.843 to 0.849 g/ml
GC>=98.5 %
View more
For analysis
RUO – Research Use Only