n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA160780010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
CAS110-54-3
CAS Min %99.0
ColorColorless to Yellow
View more
For analysis
RUO – Research Use Only