L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA160651000
Price (MYR)
525.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
525.00
Add to cart
Chemical Identifiers
CAS58-08-2
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Residue on ignition0.1%, max
Loss on Drying0.5%, max
FormPowder
Assay (Titration)>/= 99.6% ( on dry basis)