L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA160650250
Price (MYR)
343.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Specifications
Chemical Name or MaterialL(-)-Borneol
Name Note98%
CAS464-45-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.