1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA160511000
Price (MYR)
279.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
279.00
Add to cart
Chemical Identifiers
CAS12179-02-1
IUPAC Namesodium hydrate hydroxide
Molecular FormulaH3NaO2
InChI KeyJBJWASZNUJCEKT-UHFFFAOYSA-M
SMILESO.[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.