4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals
CAS: 74-11-3 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA159250250
Price (MYR)
242.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
242.00
Add to cart
Chemical Identifiers
CAS58-56-0
IUPAC Namehydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
Molecular FormulaC8H12ClNO3
InChI KeyZUFQODAHGAHPFQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
SolubilitySolution
pH2.4 to 3.0 (5 % in water)
Appearance (Form)Powder
Sulfated ash=<0.1 %