2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2 kg
Packaging:
View all
Plastic bottle
Catalog number FSA159241000
Price (MYR)
216.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
216.00
Add to cart
Chemical Identifiers
CAS6859-99-0
IUPAC Name(3S)-3-hydroxypiperidin-1-ium
Molecular FormulaC5H12NO
InChI KeyBIWOSRSKDCZIFM-YFKPBYRVSA-O
SMILESO[C@H]1CCC[NH2+]C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid or granules
Appearance (Color)White to cream to pale yellow
Assay (GC)≥98.0%
Water Content (Karl Fischer Titration)≤2%